Assay Details
Target Gene Details
Entrez Gene ID: | 17155 |
Gene Name: | mannosidase 1, alpha |
Gene Aliases: |
Man1a1, PCR1 |
Location: |
Chr.10:53904785-54076586 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man1a | NM_008548.4 | 12 | 2299 | NP_032574.1 |
| AK031038.1 | 12 | 2300 | BAC27225.1 | |
| AK042371.1 | 11 | 2140 | ||
| AK043932.1 | 12 | 2240 | ||
| AK154820.1 | 12 | 2241 | BAE32852.1 | |
| AK171841.1 | 12 | 2242 | ||
| AK191835.1 | 2 | 113 | ||
| AK192972.1 | 2 | 94 | ||
| AK196316.1 | 2 | 117 | ||
| AK196883.1 | 2 | 193 | ||
| AK197437.1 | 2 | 193 | ||
| AK201818.1 | 2 | 193 | ||
| AK204950.1 | 2 | 90 | ||
| AK211829.1 | 2 | 193 | ||
| BC015265.1 | 12 | 2200 | AAH15265.1 | |
| U04299.1 | 11 | 1903 | AAA17747.1 |
More Information
Set Membership: |
|

Genomic Map