Assay Details
Target Gene Details
Entrez Gene ID: | 17155 |
Gene Name: | mannosidase 1, alpha |
Gene Aliases: |
Man1a1, PCR1 |
Location: |
Chr.10:53904785-54076586 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man1a | NM_008548.4 | NP_032574.1 | ||
| AK028996.1 | 6 | 1422 | ||
| AK031038.1 | BAC27225.1 | |||
| AK042371.1 | ||||
| AK043932.1 | ||||
| AK154820.1 | BAE32852.1 | |||
| AK171841.1 | ||||
| AK172667.1 | 6 | 1449 | ||
| BC015265.1 | AAH15265.1 | |||
| U04299.1 | AAA17747.1 |
More Information
Set Membership: |
|

Genomic Map