Assay Details
Target Gene Details
Entrez Gene ID: | 17938 |
Gene Name: | nascent polypeptide-associated complex alpha polypeptide |
Gene Aliases: |
AL022831, AL024382, Gm1878, mKIAA0363, skNAC |
Location: |
Chr.10:128035346-128048637 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naca | NM_001113199.1 | NP_001106670.1 | ||
| NM_001282976.1 | NP_001269905.1 | |||
| NM_013608.3 | NP_038636.2 | |||
| AK146149.1 | BAE26934.1 | |||
| AK146197.1 | BAE26971.1 | |||
| AK151928.1 | BAE30804.1 | |||
| AK161678.1 | BAE36526.1 | |||
| AK168023.1 | BAE40008.1 | |||
| AK168169.1 | BAE40130.1 | |||
| AK168607.1 | BAE40474.1 | |||
| AK169432.1 | BAE41173.1 | |||
| AK169435.1 | BAE41176.1 | |||
| AL355707.1 | ||||
| BC029830.1 | AAH29830.1 | |||
| BC083340.1 | AAH83340.1 | |||
| BC099375.1 | AAH99375.1 | |||
| BI851172.1 | ||||
| BU962928.1 | ||||
| CX236470.1 | ||||
| U22151.1 | AAB80961.1 | |||
| U48364.1 | AAB18734.1 |
More Information
Set Membership: |
|

Genomic Map