Assay Details
Target Gene Details
Entrez Gene ID: | 100504234 |
Gene Name: | coiled-coil domain containing 170 |
Gene Aliases: |
Gm221 |
Location: |
Chr.10:4481416-4562374 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc170 | NM_001195672.1 | 8 | 1330 | NP_001182601.1 |

Genomic Map