Assay Details
Target Gene Details
Entrez Gene ID: | 52705 |
Gene Name: | KRR1, small subunit (SSU) processome component, homolog (yeast) |
Gene Aliases: |
2610511F02Rik, AI255219, AI428520, D10Ertd773e, Hrb2 |
Location: |
Chr.10:111972695-111988430 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krr1 | NM_178610.4 | NP_848725.2 | ||
| AK011972.1 | ||||
| AK012108.1 | ||||
| AK030560.1 | BAC27022.1 | |||
| AK032472.1 | BAC27888.1 | |||
| AK036447.1 | ||||
| AK041854.1 | BAC31083.1 | |||
| AK078627.1 | BAC37343.1 | |||
| AK146926.1 | BAE27537.1 | |||
| AK168675.1 | BAE40525.1 | |||
| BC089510.1 | AAH89510.1 | |||
| BC094236.1 | AAH94236.1 |
More Information
Set Membership: |
|

Genomic Map