Assay Details
Target Gene Details
Entrez Gene ID: | 59012 |
Gene Name: | monooxygenase, DBH-like 1 |
Gene Aliases: |
3230402N08Rik, MNCb-5203 |
Location: |
Chr.10:24223517-24302783 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Moxd1 | NM_021509.5 | NP_067484.2 | ||
| AB041606.1 | BAA95089.1 | |||
| AK014340.1 | BAB29283.1 | |||
| AK030274.1 | BAC26871.1 | |||
| AK081586.1 | BAC38265.1 | |||
| AK162030.1 | ||||
| BC025892.1 | AAH25892.1 | |||
| BC057652.1 | AAH57652.1 |
More Information
Set Membership: |
|

Genomic Map