Assay Details
Target Gene Details
Entrez Gene ID: | 80904 |
Gene Name: | deltex 3 homolog (Drosophila) |
Gene Aliases: |
Deltex3, mg310 |
Location: |
Chr.10:127190378-127196412 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3 | NM_030714.2 | NP_109639.1 | ||
| AK148444.1 | BAE28557.1 | |||
| BC099687.1 | AAH99687.1 |
Target Gene Details
Entrez Gene ID: | 100038591 |
Gene Name: | RIKEN cDNA F420014N23 gene |
Gene Aliases: |
ENSMUSG00000074658 |
Location: |
Chr.10:127195249-127202643 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F420014N23Rik | NR_045715.1 | |||
| NR_045716.1 | ||||
| AK077682.1 | ||||
| AK165234.1 | BAE38092.1 |
More Information
Set Membership: |
|

Genomic Map