Assay Details
Target Gene Details
Entrez Gene ID: | 80904 |
Gene Name: | deltex 3 homolog (Drosophila) |
Gene Aliases: |
Deltex3, mg310 |
Location: |
Chr.10:127190378-127196412 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3 | NM_030714.2 | 5 | 1182 | NP_109639.1 |
| AB015425.1 | 4 | 1164 | BAB18942.1 | |
| AK148444.1 | 5 | 1201 | BAE28557.1 | |
| AK180250.1 | ||||
| AK180771.1 | ||||
| AK181396.1 | ||||
| AK183648.1 | ||||
| AK183862.1 | ||||
| AK189235.1 | ||||
| AK195108.1 | ||||
| AK197247.1 | ||||
| AK205640.1 | ||||
| AK207039.1 | ||||
| AK209833.1 | ||||
| AK218545.1 | ||||
| AK220046.1 | ||||
| BC099687.1 | 5 | 1182 | AAH99687.1 | |
| BC138304.1 | 4 | 1061 | ||
| BC157899.1 | 4 | 1063 |
More Information
Set Membership: |
|

Genomic Map