Assay Details
Target Gene Details
Entrez Gene ID: | 66847 |
Gene Name: | histidine triad nucleotide binding protein 3 |
Gene Aliases: |
0610010I17Rik, AV009015, HINT-3, HINT-4, HINT4 |
Location: |
Chr.10:30608207-30629401 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hint3 | NM_025798.3 | NP_080074.1 | ||
| AK002482.1 | BAB22134.1 | |||
| AK007758.1 | BAB25237.1 | |||
| AK027974.1 | BAC25684.1 | |||
| AK172366.1 | 1 | 123 | ||
| AY040768.1 | AAK94778.1 | |||
| BC025065.1 | AAH25065.1 |
More Information
Set Membership: |
|

Genomic Map