Assay Details
Target Gene Details
Entrez Gene ID: | 15275 |
Gene Name: | hexokinase 1 |
Gene Aliases: |
BB404130, Hk-1, Hk1-s, dea, mHk1-s |
Location: |
Chr.10:62268855-62379920 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hk1 | NM_001146100.1 | NP_001139572.1 | ||
| NM_010438.3 | NP_034568.2 | |||
| AK143630.1 | BAE25472.1 | |||
| AK149749.1 | BAE29060.1 | |||
| AK161466.1 | BAE36411.1 | |||
| AK167468.1 | BAE39552.1 | |||
| BC072628.1 | AAH72628.1 | |||
| J05277.1 | AAA37804.1 | |||
| L16949.1 | AAB57759.1 |
More Information
Set Membership: |
|

Genomic Map