Assay Details
Target Gene Details
Entrez Gene ID: | 56307 |
Gene Name: | methionine aminopeptidase 2 |
Gene Aliases: |
4930584B20Rik, A930035J23Rik, AI047573, AL024412, AU014659, Amp2, Mnpep, p67, p67eIF2 |
Location: |
Chr.10:93858489-93897090 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap2 | NM_019648.3 | NP_062622.1 | ||
| AB003144.1 | BAA19789.1 | |||
| AF434712.1 | AAL30407.1 | |||
| AK076804.1 | BAC36488.1 | |||
| AK090323.1 | ||||
| AK147097.1 | BAE27673.1 | |||
| AK159650.1 | ||||
| AK163318.1 | BAE37297.1 | |||
| AK165301.1 | ||||
| AK165845.1 | BAE38410.1 | |||
| AK189664.1 | ||||
| AK215323.1 | ||||
| BC002213.1 | AAH02213.1 | |||
| BC092052.1 | AAH92052.1 |
More Information
Set Membership: |
|

Genomic Map