Assay Details
Target Gene Details
Entrez Gene ID: | 15275 |
Gene Name: | hexokinase 1 |
Gene Aliases: |
BB404130, Hk-1, Hk1-s, dea, mHk1-s |
Location: |
Chr.10:62268855-62379920 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hk1 | NM_001146100.1 | 13 | 2232 | NP_001139572.1 |
| NM_010438.3 | 17 | 2175 | NP_034568.2 | |
| AK143630.1 | 13 | 2170 | BAE25472.1 | |
| AK149749.1 | 13 | 1945 | BAE29060.1 | |
| AK161466.1 | 7 | 1144 | BAE36411.1 | |
| AK167468.1 | 13 | 1988 | BAE39552.1 | |
| AK197158.1 | 1 | 34 | ||
| BC072628.1 | 13 | 2353 | AAH72628.1 | |
| J05277.1 | 13 | 1890 | AAA37804.1 | |
| L16949.1 | 17 | 2154 | AAB57759.1 |
More Information
Set Membership: |
|

Genomic Map