Assay Details
Target Gene Details
Entrez Gene ID: | 15275 |
Gene Name: | hexokinase 1 |
Gene Aliases: |
BB404130, Hk-1, Hk1-s, dea, mHk1-s |
Location: |
Chr.10:62268855-62379920 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hk1 | NM_001146100.1 | 2 | 498 | NP_001139572.1 |
| NM_010438.3 | 5 | 372 | NP_034568.2 | |
| AK143630.1 | 2 | 436 | BAE25472.1 | |
| AK149749.1 | 2 | 211 | BAE29060.1 | |
| AK165237.1 | 2 | 298 | ||
| AK167468.1 | 2 | 254 | BAE39552.1 | |
| AK214520.1 | ||||
| BC072628.1 | 2 | 619 | AAH72628.1 | |
| EU430521.1 | ||||
| EU430522.1 | ||||
| EU430523.1 | ||||
| EU430524.1 | ||||
| EU430525.1 | ||||
| EU430526.1 | ||||
| EU430527.1 | ||||
| EU430528.1 | ||||
| EU430529.1 | ||||
| EU430530.1 | ||||
| EU430531.1 | ||||
| EU430532.1 | ||||
| EU430533.1 | ||||
| J05277.1 | 2 | 156 | AAA37804.1 | |
| L16949.1 | 5 | 351 | AAB57759.1 |
More Information
Set Membership: |
|

Genomic Map