Assay Details
Target Gene Details
Entrez Gene ID: | 216136 |
Gene Name: | ilvB (bacterial acetolactate synthase)-like |
Gene Aliases: |
5830463I21, AI415009 |
Location: |
Chr.10:78574345-78584502 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ilvbl | NM_173751.4 | 12 | 1556 | NP_776112.1 |
| AK030933.1 | ||||
| AK087956.1 | 11 | 1430 | BAC40057.1 | |
| AK146075.1 | 11 | 1484 | BAE26879.1 | |
| AK169899.1 | 11 | 1482 | ||
| AK184668.1 | 3 | 262 | ||
| AK218076.1 | 1 | 106 | ||
| BC056459.1 | 12 | 1514 | AAH56459.1 |
More Information
Set Membership: |
|

Genomic Map