Assay Details
Target Gene Details
Entrez Gene ID: | 260315 |
Gene Name: | neuron navigator 3 |
Gene Aliases: |
4732483H20Rik, 9630020C08Rik, POMFIL1, Pomfil1p, mKIAA0938, unc53H3 |
Location: |
Chr.10:109681573-110000391 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nav3 | NM_001081035.1 | 33 | 6113 | NP_001074504.1 |
| AK029036.1 | 1 | 85 | ||
| AK122404.1 | 22 | 3923 | BAC65686.1 |

Genomic Map