Assay Details
Target Gene Details
Entrez Gene ID: | 216344 |
Gene Name: | RAB21, member RAS oncogene family |
Gene Aliases: |
9630024B22 |
Location: |
Chr.10:115289862-115316026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab21 | NM_024454.1 | NP_077774.1 | ||
| AB232615.1 | BAF02877.1 | |||
| AK035980.1 | ||||
| AK172891.1 | BAD32169.1 | |||
| BC055042.1 | AAH55042.1 |
More Information
Set Membership: |
|

Genomic Map