Assay Details
Target Gene Details
Entrez Gene ID: | 76877 |
Gene Name: | RAB36, member RAS oncogene family |
Gene Aliases: |
6330593L16Rik, AW047545 |
Location: |
Chr.10:75036895-75054748 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab36 | NM_029781.3 | 1 | 71 | NP_084057.1 |
| AK018269.1 | 1 | 77 | BAB31140.1 | |
| AK038445.1 | 1 | 72 | BAC30001.1 | |
| AK043946.1 | 1 | 95 | BAE20645.1 | |
| AK087892.1 | 1 | 71 | ||
| AK163455.1 | 1 | 73 | ||
| BC119094.1 | 1 | 46 | ||
| BC119098.1 | 1 | 46 |

Genomic Map