Assay Details
Target Gene Details
Entrez Gene ID: | 21337 |
Gene Name: | tachykinin receptor 2 |
Gene Aliases: |
Nk2r, Skr, Tac2r |
Location: |
Chr.10:62252438-62265990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tacr2 | NM_009314.4 | NP_033340.3 | ||
| AK033443.1 | BAC28290.1 | |||
| BC106959.1 | AAI06960.1 | |||
| X62933.1 | CAA44706.1 |
More Information
Set Membership: |
|

Genomic Map