Assay Details
Target Gene Details
Entrez Gene ID: | 21886 |
Gene Name: | transducin-like enhancer of split 2, homolog of Drosophila E(spl) |
Gene Aliases: |
Grg2, mKIAA4188 |
Location: |
Chr.10:81574739-81592713 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle2 | NM_001252401.1 | NP_001239330.1 | ||
| NM_019725.2 | NP_062699.1 | |||
| AF145958.2 | AAD37694.1 | |||
| AK144581.1 | BAE25947.1 | |||
| AK160999.1 | 5 | 1145 | ||
| AK220533.1 | BAD90529.1 | |||
| BC051955.1 | AAH51955.1 | |||
| BC071219.1 | AAH71219.1 |
More Information
Set Membership: |
|

Genomic Map