Assay Details
Target Gene Details
Entrez Gene ID: | 21886 |
Gene Name: | transducin-like enhancer of split 2, homolog of Drosophila E(spl) |
Gene Aliases: |
Grg2, mKIAA4188 |
Location: |
Chr.10:81574739-81592713 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle2 |
Target Gene Details
Entrez Gene ID: | 114606 |
Gene Name: | transducin-like enhancer of split 6, homolog of Drosophila E(spl) |
Gene Aliases: |
1810057E06Rik, Grg6 |
Location: |
Chr.10:81590904-81601190 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle6 | NM_053254.2 | NP_444484.1 | ||
| AB221587.1 | ||||
| AF145957.2 | AAD37693.1 | |||
| AK007880.1 | ||||
| AK154391.1 | ||||
| BC048832.1 | AAH48832.1 |
More Information
Set Membership: |
|

Genomic Map