Assay Details
Target Gene Details
Entrez Gene ID: | 76877 |
Gene Name: | RAB36, member RAS oncogene family |
Gene Aliases: |
6330593L16Rik, AW047545 |
Location: |
Chr.10:75036895-75054748 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab36 | NM_029781.3 | NP_084057.1 | ||
| AB232632.1 | BAF02894.1 | |||
| AK018269.1 | BAB31140.1 | |||
| AK038445.1 | BAC30001.1 | |||
| AK043946.1 | BAE20645.1 | |||
| AK046113.1 | BAE20653.1 | |||
| AK142025.1 | BAE24917.1 | |||
| BC119094.1 | ||||
| BC119098.1 |

Genomic Map