Assay Details
Target Gene Details
Entrez Gene ID: | 216363 |
Gene Name: | RAB3A interacting protein |
Gene Aliases: |
B230311A06, Gtpat12, Rabin3 |
Location: |
Chr.10:116905780-116950448 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3ip | NM_001003950.2 | NP_001003950.1 | ||
| AK152122.1 | ||||
| AK178592.1 | ||||
| BC080289.1 | AAH80289.1 | |||
| S79539.1 | AAB35170.1 |
More Information
Set Membership: |
|

Genomic Map