Assay Details
Target Gene Details
Entrez Gene ID: | 216441 |
Gene Name: | solute carrier family 26, member 10 |
Gene Aliases: |
C78409, D630010I07 |
Location: |
Chr.10:127172313-127180709 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a10 | NM_177615.3 | 17 | 2010 | NP_808283.2 |
More Information
Set Membership: |
|

Genomic Map