Assay Details
Target Gene Details
Entrez Gene ID: | 74694 |
Gene Name: | TBC1 domain family, member 30 |
Gene Aliases: |
4930505D03Rik |
Location: |
Chr.10:121263819-121311189 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d30 | NM_029057.1 | NP_083333.1 | ||
| AK044521.1 | BAC31963.1 | |||
| AK049989.1 | ||||
| AK173079.1 | BAD32357.1 | |||
| BC079587.1 | AAH79587.1 | |||
| BC138285.1 | ||||
| BC138286.1 |
More Information
Set Membership: |
|

Genomic Map