Assay Details
Target Gene Details
Entrez Gene ID: | 74694 |
Gene Name: | TBC1 domain family, member 30 |
Gene Aliases: |
4930505D03Rik |
Location: |
Chr.10:121263819-121311189 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d30 | NM_029057.1 | 6 | 1068 | NP_083333.1 |
| AK044521.1 | 6 | 875 | BAC31963.1 | |
| AK049989.1 | 6 | 901 | ||
| AK173079.1 | 6 | 831 | BAD32357.1 | |
| BC079587.1 | 3 | 379 | AAH79587.1 | |
| BC138285.1 | 6 | 1275 | ||
| BC138286.1 | 6 | 1275 |
More Information
Set Membership: |
|

Genomic Map