Assay Details
Target Gene Details
Entrez Gene ID: | 320995 |
Gene Name: | regulatory factor X, 6 |
Gene Aliases: |
4930572O07Rik, Rfxdc1 |
Location: |
Chr.10:51677752-51730432 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx6 | NM_001159389.1 | 18 | 2542 | NP_001152861.1 |
| NM_177306.3 | 15 | 2173 | NP_796280.1 | |
| AK029809.1 | 15 | 2174 | BAC26627.1 | |
| BC132104.1 | 13 | 1856 | ||
| BC132294.1 | 13 | 1856 | ||
| FJ804414.1 | 17 | 2390 |
More Information
Set Membership: |
|

Genomic Map