Assay Details
Target Gene Details
Entrez Gene ID: | 71279 |
Gene Name: | solute carrier family 29 (nucleoside transporters), member 3 |
Gene Aliases: |
4933435C21Rik, AW987637, Ent3 |
Location: |
Chr.10:60712072-60752850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc29a3 | NM_023596.3 | NP_076085.1 | ||
| AF326986.1 | AAK00957.1 | |||
| AK034133.1 | BAC28600.1 | |||
| AK051317.1 | ||||
| AK157868.1 | ||||
| BC137864.1 | ||||
| CV557129.1 |
More Information
Set Membership: |
|

Genomic Map