Assay Details
Target Gene Details
Entrez Gene ID: | 78808 |
Gene Name: | syntaxin binding protein 5 (tomosyn) |
Gene Aliases: |
0710001E20Rik, 4930565N16Rik, AW742610, LGL3, mKIAA1006 |
Location: |
Chr.10:9755547-9901227 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp5 | NM_001081344.2 | 25 | 3600 | NP_001074813.2 |
| AF516607.1 | 21 | 2679 | AAM61881.1 | |
| AK019788.1 | 8 | 1341 | BAB31853.1 | |
| AK029982.1 | 6 | 3351 | ||
| AK122417.1 | 4 | 679 | BAC65699.1 |
More Information
Set Membership: |
|

Genomic Map