Assay Details
Target Gene Details
Entrez Gene ID: | 28240 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 2 |
Gene Aliases: |
9830168K16Rik, C79133, LTRPC2, TRPC7, Trp7, Trrp7 |
Location: |
Chr.10:77907722-77969906 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpm2 | NM_138301.2 | NP_612174.2 | ||
| AB166747.1 | BAD83707.1 | |||
| AJ344343.1 | CAC69081.1 | |||
| AJ878415.1 | CAI47592.1 | |||
| AK134735.1 | BAE22263.1 | |||
| AK136905.1 | ||||
| BC141391.1 |
More Information
Set Membership: |
|

Genomic Map