Assay Details
Target Gene Details
Entrez Gene ID: | 320995 |
Gene Name: | regulatory factor X, 6 |
Gene Aliases: |
4930572O07Rik, Rfxdc1 |
Location: |
Chr.10:51677752-51730432 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx6 | NM_001159389.1 | NP_001152861.1 | ||
| NM_177306.3 | NP_796280.1 | |||
| AK029809.1 | BAC26627.1 | |||
| AK049380.1 | BAC33722.1 | |||
| FJ804414.1 |
More Information
Set Membership: |
|

Genomic Map