Assay Details
Target Gene Details
Entrez Gene ID: | 211389 |
Gene Name: | sulfite oxidase |
Gene Aliases: |
SO |
Location: |
Chr.10:128669887-128673918 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suox | NM_173733.3 | NP_776094.2 | ||
| AK149422.1 | BAE28865.1 | |||
| AK154608.1 | BAE32710.1 |
More Information
Set Membership: |
|

Genomic Map