Assay Details
Target Gene Details
Entrez Gene ID: | 21390 |
Gene Name: | thromboxane A2 receptor |
Gene Aliases: |
TP, TXA2-R |
Location: |
Chr.10:81328259-81335184 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbxa2r | NM_001277265.1 | 1 | 159 | NP_001264194.1 |
| NM_009325.4 | 1 | 159 | NP_033351.1 | |
| AK158477.1 | 1 | 132 | BAE34532.1 | |
| BC089611.1 | 1 | 132 | AAH89611.1 | |
| D10849.1 | BAA01622.1 |
More Information
Set Membership: |
|

Genomic Map