Assay Details
Target Gene Details
Entrez Gene ID: | 215449 |
Gene Name: | RAS related protein 1b |
Gene Aliases: |
2810443E11Rik |
Location: |
Chr.10:117814597-117845974 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap1b | NM_024457.2 | NP_077777.1 | ||
| AK143910.1 | BAE25597.1 | |||
| AK153870.1 | BAE32223.1 | |||
| AK163242.1 | BAE37252.1 | |||
| AK167548.1 | BAE39614.1 | |||
| AK178034.1 | ||||
| AK196093.1 | ||||
| AK199293.1 | ||||
| AK218552.1 | ||||
| BC033382.1 | AAH33382.2 | |||
| BC052480.1 | AAH52480.1 | |||
| BC094066.1 | AAH94066.1 | |||
| BC147390.1 | ||||
| BC147391.1 | ||||
| BC147700.1 | ||||
| BC147713.1 | ||||
| M79314.1 | AAK14823.1 |
More Information
Set Membership: |
|

Genomic Map