Assay Details
Target Gene Details
Entrez Gene ID: | 216161 |
Gene Name: | strawberry notch homolog 2 (Drosophila) |
Gene Aliases: |
BC019206, F730040C21, Sno, Stno |
Location: |
Chr.10:80056997-80106108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sbno2 | NM_183426.1 | NP_906271.1 | ||
| AJ512611.1 | CAD54758.1 | |||
| AK149806.1 | BAE29097.1 | |||
| AK155462.1 | ||||
| AK155674.1 | BAE33382.1 | |||
| BC056369.1 | AAH56369.1 |

Genomic Map