Assay Details
Target Gene Details
Entrez Gene ID: | 192678 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 3 |
Gene Aliases: |
AW212023, AW322379 |
Location: |
Chr.10:121410350-121476250 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf3 | NM_138956.3 | 3 | 472 | NP_620406.1 |
| AF332864.1 | 3 | 483 | AAK06849.1 | |
| AK155495.1 | 3 | 499 | BAE33293.1 | |
| AK167627.1 | 3 | 305 | BAE39678.1 | |
| BC011511.1 | 3 | 472 | AAH11511.1 | |
| BC055023.1 | 3 | 527 | AAH55023.1 | |
| BC071204.1 | 3 | 480 | AAH71204.1 |
More Information
Set Membership: |
|

Genomic Map