Assay Details
Target Gene Details
Entrez Gene ID: | 216161 |
Gene Name: | strawberry notch homolog 2 (Drosophila) |
Gene Aliases: |
BC019206, F730040C21, Sno, Stno |
Location: |
Chr.10:80056997-80106108 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sbno2 | NM_183426.1 | 28 | 3344 | NP_906271.1 |
| AJ512611.1 | 29 | 3497 | CAD54758.1 | |
| AK149806.1 | 28 | 3365 | BAE29097.1 | |
| AK155462.1 | 28 | 3339 | ||
| AK155674.1 | 28 | 3338 | BAE33382.1 | |
| BC056369.1 | 28 | 3344 | AAH56369.1 |

Genomic Map