Assay Details
Target Gene Details
Entrez Gene ID: | 117600 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 1 |
Gene Aliases: |
4930572H05Rik, Arhgap13 |
Location: |
Chr.10:121780991-122047315 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srgap1 | NM_001081037.2 | NP_001074506.1 | ||
| NM_001242411.1 | NP_001229340.1 | |||
| AY057898.1 | AAL27030.1 | |||
| BC120892.1 | ||||
| BC120893.1 | ||||
| BU706317.1 | ||||
| CX569081.1 |
More Information
Set Membership: |
|

Genomic Map