Assay Details
Target Gene Details
Entrez Gene ID: | 117600 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 1 |
Gene Aliases: |
4930572H05Rik, Arhgap13 |
Location: |
Chr.10:121780991-122047315 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srgap1 | NM_001081037.2 | 17 | 2197 | NP_001074506.1 |
| NM_001242411.1 | 17 | 2128 | NP_001229340.1 | |
| AK016279.1 | 2 | 374 | ||
| AK016805.1 | 2 | 374 | ||
| AY057898.1 | 10 | 959 | AAL27030.1 | |
| BC120892.1 | 17 | 2121 | ||
| BC120893.1 | 17 | 2121 | ||
| BU704234.1 | 2 | 248 | ||
| BU706317.1 | 6 | 569 | ||
| CX569081.1 |
More Information
Set Membership: |
|

Genomic Map