Assay Details
Target Gene Details
Entrez Gene ID: | 71365 |
Gene Name: | prenyl (solanesyl) diphosphate synthase, subunit 2 |
Gene Aliases: |
5430420P03Rik, Gm60, Plmp, kd, mDLP1 |
Location: |
Chr.10:43220310-43464882 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdss2 | NM_001168289.1 | 3 | 809 | NP_001161761.1 |
| NM_027772.2 | 3 | 809 | NP_082048.2 | |
| AB210840.1 | 3 | 533 | BAE48218.1 | |
| AK017329.1 | 3 | 779 | BAB30693.1 | |
| AK140091.1 | 3 | 696 | BAE24234.1 | |
| AK165732.1 | 3 | 808 | ||
| BC147693.1 | 3 | 586 | ||
| BC147694.1 | 3 | 586 | ||
| BC147752.1 | 3 | 559 | ||
| BC147753.1 | 3 | 559 |

Genomic Map