Assay Details
Target Gene Details
Entrez Gene ID: | 71365 |
Gene Name: | prenyl (solanesyl) diphosphate synthase, subunit 2 |
Gene Aliases: |
5430420P03Rik, Gm60, Plmp, kd, mDLP1 |
Location: |
Chr.10:43220310-43464882 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdss2 | NM_001168289.1 | NP_001161761.1 | ||
| NM_027772.2 | NP_082048.2 | |||
| AB210840.1 | BAE48218.1 | |||
| AK140091.1 | BAE24234.1 | |||
| BC147693.1 | ||||
| BC147694.1 |
More Information
Set Membership: |
|

Genomic Map