Assay Details
Target Gene Details
Entrez Gene ID: | 215890 |
Gene Name: | clavesin 2 |
Gene Aliases: |
A330019N05Rik, Rlbp1l2 |
Location: |
Chr.10:33512286-33624905 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clvs2 | NM_175448.3 | 5 | 1380 | NP_780657.1 |
| AK039308.1 | 5 | 1381 | BAC30312.1 | |
| AK049896.1 | 5 | 941 | BAC33976.1 | |
| AK049988.1 | 5 | 949 | ||
| AK139409.1 | 5 | 1455 | BAE23998.1 | |
| AK139868.1 | 5 | 1433 | ||
| AK169009.1 | 5 | 1550 | BAE40806.1 | |
| BC058539.1 | 5 | 1387 | AAH58539.1 |
More Information
Set Membership: |
|

Genomic Map