Assay Details
Target Gene Details
Entrez Gene ID: | 213326 |
Gene Name: | SCY1-like 2 (S. cerevisiae) |
Gene Aliases: |
AU015688, AW536445, BC030932, CVAK104, D10Ertd802e |
Location: |
Chr.10:89638720-89686292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scyl2 | NM_198021.2 | NP_932138.1 | ||
| AK139747.1 | BAE24123.1 | |||
| AK139761.1 | ||||
| BC042443.1 | AAH42443.1 | |||
| BC042770.1 | AAH42770.1 |
More Information
Set Membership: |
|

Genomic Map