Assay Details
Target Gene Details
Entrez Gene ID: | 71924 |
Gene Name: | epsilon-tubulin 1 |
Gene Aliases: |
2310061K05Rik, AI551343, Tube |
Location: |
Chr.10:39133979-39151058 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tube1 | NM_028006.2 | 8 | 776 | NP_082282.1 |
| AK010005.1 | 8 | 770 | BAB26636.1 | |
| AK040492.1 | 1 | 1692 | ||
| AK041416.1 | 8 | 741 | ||
| AK087403.1 | ||||
| AK148039.1 | 8 | 776 | BAE28305.1 | |
| BC062179.1 | 8 | 773 | AAH62179.1 |
More Information
Set Membership: |
|

Genomic Map