Assay Details
Target Gene Details
Entrez Gene ID: | 14533 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 1 |
Gene Aliases: |
AI839753, BLOS1, Gcn5l1 |
Location: |
Chr.10:128919220-128923547 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s1 |
Target Gene Details
Entrez Gene ID: | 19682 |
Gene Name: | retinol dehydrogenase 5 |
Gene Aliases: |
9-cis, AI987873, RDH4, cRDH |
Location: |
Chr.10:128913588-128919297 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh5 | NM_134006.4 | NP_598767.1 | ||
| AF033196.2 | AAC00492.1 | |||
| AK033063.1 | BAC28142.1 | |||
| AK135139.1 | BAE22435.1 |
More Information
Set Membership: |
|

Genomic Map