Assay Details
Target Gene Details
Entrez Gene ID: | 54712 |
Gene Name: | plexin C1 |
Gene Aliases: |
2510048K12Rik, AW742158, CD232, Vespr |
Location: |
Chr.10:94790866-94945156 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plxnc1 | NM_018797.2 | NP_061267.1 | ||
| AF190578.1 | AAF01334.1 | |||
| AK084603.1 | BAC39225.1 |
More Information
Set Membership: |
|

Genomic Map