Assay Details
Target Gene Details
Entrez Gene ID: | 18641 |
Gene Name: | phosphofructokinase, liver, B-type |
Gene Aliases: |
AA407869, ATP-PFK, PFK-L |
Location: |
Chr.10:77986947-78009796 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfkl | NM_008826.4 | 1 | 135 | NP_032852.2 |
| AK008812.1 | 1 | 89 | ||
| AK030511.1 | 1 | 90 | BAC26997.1 | |
| AK036318.1 | 1 | 68 | BAC29382.1 | |
| AK081313.1 | 1 | 86 | BAC38193.1 | |
| AK159328.1 | 1 | 80 | BAE34994.1 | |
| AK168660.1 | 1 | 90 | ||
| BC020097.1 | 1 | 89 | AAH20097.1 | |
| BY068830.1 | 1 | 136 | ||
| J03928.1 | 1 | 64 | AAA20076.1 |
More Information
Set Membership: |
|

Genomic Map