Assay Details
Target Gene Details
Entrez Gene ID: | 21929 |
Gene Name: | tumor necrosis factor, alpha-induced protein 3 |
Gene Aliases: |
A20, Tnfip3 |
Location: |
Chr.10:19000910-19015410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfaip3 | NM_001166402.1 | NP_001159874.1 | ||
| NM_009397.3 | NP_033423.3 | |||
| AK088297.1 | BAC40267.1 | |||
| AK151921.1 | BAE30799.1 | |||
| AK153238.1 | BAE31830.1 | |||
| AK171326.1 | BAE42395.1 | |||
| AK172529.1 | BAE43051.1 | |||
| BC054806.1 | AAH54806.1 | |||
| BC060221.1 | AAH60221.1 | |||
| CT010397.1 | CAJ18603.1 | |||
| U19463.1 | AAC52153.1 |
More Information
Set Membership: |
|

Genomic Map