Assay Details
Target Gene Details
Entrez Gene ID: | 18641 |
Gene Name: | phosphofructokinase, liver, B-type |
Gene Aliases: |
AA407869, ATP-PFK, PFK-L |
Location: |
Chr.10:77986947-78009796 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfkl | NM_008826.4 | NP_032852.2 | ||
| AK030511.1 | BAC26997.1 | |||
| AK030892.1 | BAC27175.1 | |||
| AK036318.1 | BAC29382.1 | |||
| AK081313.1 | BAC38193.1 | |||
| AK159328.1 | BAE34994.1 | |||
| AK168660.1 | ||||
| AK185403.1 | ||||
| AK194790.1 | ||||
| AK208650.1 | ||||
| AK209868.1 | ||||
| BC020097.1 | AAH20097.1 | |||
| J03928.1 | AAA20076.1 |
More Information
Set Membership: |
|

Genomic Map