Assay Details
Target Gene Details
Entrez Gene ID: | 18641 |
Gene Name: | phosphofructokinase, liver, B-type |
Gene Aliases: |
AA407869, ATP-PFK, PFK-L |
Location: |
Chr.10:77986947-78009796 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfkl | NM_008826.4 | 21 | 2256 | NP_032852.2 |
| AK030511.1 | 21 | 2211 | BAC26997.1 | |
| AK030892.1 | 14 | 1355 | BAC27175.1 | |
| AK036318.1 | 21 | 2189 | BAC29382.1 | |
| AK081313.1 | 21 | 2207 | BAC38193.1 | |
| AK159328.1 | 21 | 2201 | BAE34994.1 | |
| AK168660.1 | 21 | 2212 | ||
| BC020097.1 | 21 | 2210 | AAH20097.1 | |
| J03928.1 | 21 | 2185 | AAA20076.1 |
More Information
Set Membership: |
|

Genomic Map