Assay Details
Target Gene Details
Entrez Gene ID: | 19707 |
Gene Name: | RalBP1 associated Eps domain containing protein |
Gene Aliases: |
BB161292 |
Location: |
Chr.10:18055561-18125155 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reps1 | NM_001111065.1 | 1 | 86 | NP_001104535.1 |
| NM_009048.2 | 1 | 86 | NP_033074.2 | |
| BC087547.1 | AAH87547.1 | |||
| BF318674.1 | 1 | 86 |
More Information
Set Membership: |
|

Genomic Map